Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286238.1 | NP_001273167.1 | 13 | 3762 | ||
NM_001286239.1 | NP_001273168.1 | 12 | 3768 | |||
NM_019073.3 | NP_061946.1 | 13 | 3810 | |||
XM_006710700.2 | XP_006710763.1 | 11 | 3609 | |||
XM_011541606.1 | XP_011539908.1 | 11 | 3519 | |||
XM_011541607.2 | XP_011539909.1 | 10 | 3373 | |||
XM_011541608.1 | XP_011539910.1 | 12 | 3747 | |||
XM_011541609.2 | XP_011539911.1 | 11 | 3455 | |||
XM_017001510.1 | XP_016856999.1 | 10 | 3559 | |||
XM_017001511.1 | XP_016857000.1 | 12 | 3712 | |||
XM_017001512.1 | XP_016857001.1 | 14 | 3653 | |||
XM_017001513.1 | XP_016857002.1 | 11 | 3417 | |||
XM_017001514.1 | XP_016857003.1 | 14 | 3695 | |||
XM_017001515.1 | XP_016857004.1 | 12 | 3540 | |||
GenBank mRNA | BC040655.1 | - | 1 | 221 |