Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAAA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005262925.3 | XP_005262982.1 | 9 | 2496 | ||
XM_017008029.1 | XP_016863518.1 | 10 | 2525 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK025371.1 | - | 1 | 98 | Chr. 4 - 75914379 - 75916406 [-] on Build GRCh38 |