Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100287290 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK093960.1 | - | 1 | 1244 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI561634.1 | - | 3 | 304 | Chr. 3 - 158732361 - 158737497 [+] on Build GRCh38 |