Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIO3OS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC037351.1 | - | 1 | 1855 | ||
BC065701.1 | - | 3 | 629 | |||
BC111049.1 | - | 2 | 446 |