Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUGT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011534891.2 | XP_011533193.1 | 14 | 11046 | ||
XM_011534893.2 | XP_011533195.1 | 13 | 10764 | |||
XM_017020365.1 | XP_016875854.1 | 14 | 10936 | |||
XM_017020366.1 | XP_016875855.1 | 14 | 10811 | |||
XM_017020367.1 | XP_016875856.1 | 14 | 10845 | |||
XM_017020368.1 | XP_016875857.1 | 14 | 10857 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AL831999.1 | - | 2 | 2876 | Chr. 13 - 52689587 - 52700758 [+] on Build GRCh38 |