Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RUFY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC030097.1 | - | 1 | 530 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG707400.1 | - | 1 | 530 | Chr. 10 - 68354953 - 68355693 [-] on Build GRCh38 |