Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017023352.1 | XP_016878841.1 | 10 | 1177 | ||
XM_017023353.1 | XP_016878842.1 | 9 | 1098 | |||
XM_017023355.1 | XP_016878844.1 | 8 | 946 | |||
GenBank mRNA | AK123247.1 | - | 1 | 484 |