Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104387.1 | 40 | 5982 | |||
NR_104388.1 | 40 | 5973 | ||||
NR_104389.1 | 39 | 5880 | ||||
NR_104390.1 | 40 | 5886 | ||||
NR_104391.1 | 41 | 5908 | ||||
NR_104392.1 | 40 | 5902 | ||||
NR_104393.1 | 41 | 5988 | ||||
NR_104394.1 | 40 | 5859 | ||||
NR_104395.1 | 40 | 5911 | ||||
NR_104396.1 | 3 | 1347 | ||||
GenBank mRNA | AJ549245.1 | - | 2 | 885 | ||
AJ549246.2 | - | 3 | 1347 | |||
AJ549247.1 | - | 5 | 672 | |||
AJ549248.3 | - | 15 | 1807 | |||
AJ549249.2 | - | 11 | 1344 | |||
AJ549250.3 | - | 14 | 1705 | |||
AJ555148.2 | - | 15 | 1711 | |||
AJ555149.2 | - | 16 | 1733 |