Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WIPF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005257083.2 | XP_005257140.1 | 8 | 7461 | ||
XM_005257084.2 | XP_005257141.1 | 8 | 7419 | |||
XM_011524412.1 | XP_011522714.1 | 7 | 7360 | |||
XM_011524413.1 | XP_011522715.1 | 7 | 7341 | |||
GenBank mRNA | BC030561.1 | - | 1 | 1454 |