Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC43A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284498.1 | NP_001271427.1 | 15 | 6870 | ||
NM_001284499.1 | NP_001271428.1 | 10 | 6311 | |||
NM_001321364.1 | NP_001308293.1 | 15 | 6668 | |||
NM_001321365.1 | NP_001308294.1 | 15 | 6837 | |||
NM_152346.2 | NP_689559.1 | 14 | 6656 | |||
GenBank mRNA | AK056769.1 | - | 1 | 1748 |