Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204911.1 | NP_001191840.1 | 4 | 671 | ||
GenBank mRNA | AK124443.1 | - | 1 | 466 | ||
AK301494.1 | - | 4 | 621 |