Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IAH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK123278.1 | - | 1 | 693 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX644159.1 | - | 1 | 22 | Chr. 2 - 9498405 - 9499010 [+] on Build GRCh38 |