Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001194999.1 | NP_001181928.1 | 7 | 7374 | ||
NM_001195000.1 | NP_001181929.1 | 6 | 7233 | |||
NM_001195035.1 | NP_001181964.1 | 5 | 7128 | |||
NM_001310213.1 | NP_001297142.1 | 7 | 7349 | |||
NM_001531.2 | NP_001522.1 | 7 | 7509 | |||
XM_006711286.3 | XP_006711349.1 | 7 | 7468 | |||
XM_011509463.2 | XP_011507765.1 | 6 | 7281 | |||
GenBank mRNA | AK056155.1 | - | 1 | 2150 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX114137.1 | - | 1 | 453 | Chr. 1 - 181061135 - 181061726 [+] on Build GRCh38 |