Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206836.1 | NP_001193765.1 | 5 | 3871 | ||
NM_004663.4 | NP_004654.1 | 5 | 3966 | |||
GenBank mRNA | BC047114.1 | - | 1 | 258 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CN260553.1 | - | 1 | 147 | Chr. 15 - 65890882 - 65891531 [+] on Build GRCh38 |