Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPBGL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195528.1 | NP_001182457.1 | 1 | 2695 | ||
GenBank mRNA | BC007804.2 | - | 1 | 1405 | ||
N54218.1 | - | 1 | 121 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BP395974.1 | - | 1 | 259 | Chr. 11 - 75243342 - 75243683 [+] on Build GRCh38 |