Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005248218.4 | XP_005248275.1 | 5 | 4030 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK001995.1 | - | 1 | 2048 | Chr. 4 - 13456451 - 13458546 [-] on Build GRCh38 |