Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006722639.2 | XP_006722702.1 | 1 | 544 | ||
XM_017026249.1 | XP_016881738.1 | 1 | 544 | |||
XM_017026250.1 | XP_016881739.1 | 1 | 544 | |||
XM_017026251.1 | XP_016881740.1 | 1 | 544 | |||
XM_017026252.1 | XP_016881741.1 | 1 | 544 | |||
GenBank mRNA | AK055767.1 | - | 1 | 544 |