Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCF2L-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034002.1 | 1 | 674 | |||
GenBank mRNA | BC064982.1 | - | 675 | |||
BC080517.1 | - | 675 |