Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCUN1D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308101.1 | NP_001295030.1 | 7 | 6732 | ||
NM_020640.3 | NP_065691.2 | 7 | 6714 | |||
XM_011512912.1 | XP_011511214.1 | 7 | 2049 | |||
XM_011512914.1 | XP_011511216.1 | 7 | 2088 | |||
XM_011512915.2 | XP_011511217.1 | 8 | 2610 | |||
XM_011512916.2 | XP_011511218.1 | 7 | 2538 | |||
XM_017006633.1 | XP_016862122.1 | 7 | 1860 | |||
XM_017006634.1 | XP_016862123.1 | 7 | 2054 | |||
GenBank mRNA | AK095773.1 | - | 1 | 1418 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY994955.1 | - | 1 | 98 | Chr. 3 - 182938666 - 182939353 [-] on Build GRCh38 |