Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POU6F2-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK023033.1 | - | 4 | 425 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AU129608.1 | - | 4 | 425 | Chr. 7 - 38984271 - 39013567 [-] on Build GRCh38 |