Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AHSA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011532541.2 | XP_011530843.1 | 8 | 2126 | ||
XM_011532543.2 | XP_011530845.1 | 9 | 2410 | |||
XM_017003378.1 | XP_016858867.1 | 9 | 2085 | |||
GenBank mRNA | AK091998.1 | - | 1 | 1302 |
Gene Symbol: USP34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AW293998.1 | - | 1 | 142 |