Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305043.1 | NP_001291972.1 | 8 | 5318 | ||
NM_001305049.1 | NP_001291978.1 | 8 | 5097 | |||
NM_001305050.1 | NP_001291979.1 | 6 | 5117 | |||
NM_001305051.1 | NP_001291980.1 | 8 | 5094 | |||
NM_001305052.1 | NP_001291981.1 | 7 | 4988 | |||
NM_004872.4 | NP_004863.2 | 8 | 5315 | |||
GenBank mRNA | AK057652.1 | - | 1 | 443 |