Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COLCA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017017197.1 | XP_016872686.1 | 1 | 356 | ||
GenBank mRNA | AB096246.1 | - | 1 | 801 |