Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016116.2 | NP_057200.1 | 5 | 1419 | ||
GenBank mRNA | AK096449.1 | - | 1 | 103 |