Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005267510.1 | XP_005267567.1 | 7 | 5259 | ||
GenBank mRNA | AK025898.1 | - | 1 | 1871 | ||
BC064901.1 | - | 5 | 2103 |