Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128166.2 | NP_001121638.1 | 16 | 4902 | ||
NM_001128167.2 | NP_001121639.1 | 15 | 4829 | |||
NM_001128168.2 | NP_001121640.1 | 20 | 5151 | |||
NM_001128172.2 | NP_001121644.1 | 15 | 4504 | |||
NM_001128173.2 | NP_001121645.1 | 19 | 5088 | |||
NM_001324325.1 | NP_001311254.1 | 16 | 4980 | |||
NM_001324326.1 | NP_001311255.1 | 16 | 4825 | |||
NM_001324327.1 | NP_001311256.1 | 18 | 4937 | |||
NM_001324328.1 | NP_001311257.1 | 17 | 4870 | |||
NM_001324329.1 | NP_001311258.1 | 19 | 5085 | |||
NM_001324330.1 | NP_001311259.1 | 16 | 4977 | |||
NM_001324331.1 | NP_001311260.1 | 17 | 4892 | |||
NM_001324332.1 | NP_001311261.1 | 18 | 5043 | |||
NM_001324333.1 | NP_001311262.1 | 18 | 5018 | |||
NM_001324334.1 | NP_001311263.1 | 17 | 4973 | |||
NM_002578.4 | NP_002569.1 | 18 | 5040 | |||
NR_136747.1 | 18 | 5067 | ||||
NR_136748.1 | 16 | 4702 | ||||
XM_017029557.1 | XP_016885046.1 | 17 | 4939 | |||
XM_017029561.1 | XP_016885050.1 | 18 | 5042 | |||
XM_017029563.1 | XP_016885052.1 | 20 | 5143 | |||
GenBank mRNA | AK124702.1 | - | 1 | 618 |