Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHISA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173461.1 | NP_001166932.1 | 4 | 5718 | ||
NM_001173462.1 | NP_001166933.1 | 5 | 5814 | |||
NM_207386.3 | NP_997269.2 | 6 | 5871 | |||
XM_017024618.1 | XP_016880107.1 | 5 | 5775 | |||
GenBank mRNA | AK094349.1 | - | 1 | 762 | ||
AK095052.1 | - | 1 | 1364 | |||
BC105704.1 | - | 1 | 520 | |||
BC105705.1 | - | 1 | 520 | |||
BC105706.1 | - | 1 | 519 | |||
BC105707.1 | - | 1 | 520 |