Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA1614-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK026359.1 | - | 2 | 1042 |
Gene Symbol: STX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011509058.1 | XP_011507360.1 | 8 | 1961 | ||
XM_011509059.1 | XP_011507361.1 | 7 | 1736 | |||
XM_017000006.1 | XP_016855495.1 | 6 | 1570 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000415647.1 | - | 2 | 1042 | Chr. 1 - 180949699 - 180954887 [-] on Build GRCh38 |