Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005273032.3 | XP_005273089.1 | 6 | 6623 | ||
XM_011544041.2 | XP_011542343.1 | 7 | 6722 | |||
XM_011544043.2 | XP_011542345.1 | 9 | 7055 | |||
XM_011544044.2 | XP_011542346.1 | 8 | 6826 | |||
XM_011544045.2 | XP_011542347.1 | 8 | 6951 | |||
XM_011544046.2 | XP_011542348.1 | 6 | 6659 | |||
XM_011544047.2 | XP_011542349.1 | 6 | 6595 | |||
XM_017000152.1 | XP_016855641.1 | 8 | 9648 | |||
XM_017000153.1 | XP_016855642.1 | 7 | 6726 | |||
XM_017000154.1 | XP_016855643.1 | 6 | 6420 | |||
XM_017000155.1 | XP_016855644.1 | 7 | 6763 | |||
XM_017000156.1 | XP_016855645.1 | 7 | 6727 | |||
XM_017000157.1 | XP_016855646.1 | 5 | 6328 | |||
XM_017000158.1 | XP_016855647.1 | 5 | 6560 | |||
XM_017000159.1 | XP_016855648.1 | 5 | 6532 | |||
XM_017000160.1 | XP_016855649.1 | 4 | 7382 | |||
XM_017000161.1 | XP_016855650.1 | 6 | 6664 | |||
XM_017000162.1 | XP_016855651.1 | 5 | 6496 | |||
XM_017000163.1 | XP_016855652.1 | 4 | 10358 | |||
GenBank mRNA | AK095715.1 | - | 1 | 1328 |