Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258312.1 | NP_001245241.1 | 16 | 3435 | ||
NM_005019.4 | NP_005010.2 | 15 | 3450 | |||
XM_017004295.1 | XP_016859784.1 | 16 | 3455 | |||
XM_017004296.1 | XP_016859785.1 | 16 | 3377 | |||
XM_017004297.1 | XP_016859786.1 | 16 | 3318 | |||
XM_017004298.1 | XP_016859787.1 | 16 | 3500 | |||
XM_017004302.1 | XP_016859791.1 | 15 | 3582 | |||
GenBank mRNA | BC047057.1 | - | 1 | 125 |