Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253384.1 | NP_001240313.1 | 5 | 1430 | ||
GenBank mRNA | BC081565.1 | - | 5 | 1095 | 5458912 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK055501.1 | - | 1 | 33 | Chr. 3 - 23922255 - 23924373 [+] on Build GRCh38 |