Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011528330.2 | XP_011526632.2 | 7 | 1051 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC017563.1 | - | 406 | Chr. - - [-] on Build |