Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017023733.1 | XP_016879222.1 | 1 | 1686 | ||
GenBank mRNA | AK127550.1 | - | 1 | 1686 |