Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005255571.2 | XP_005255628.1 | 10 | 6295 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK097189.1 | - | 1 | 920 | Chr. 16 - 8650168 - 8652458 [+] on Build GRCh38 |