Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTLL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006712875.2 | XP_006712938.1 | 20 | 5776 | ||
XM_017005387.1 | XP_016860876.1 | 20 | 5954 | |||
XM_017005388.1 | XP_016860877.1 | 21 | 5841 | |||
XM_017005389.1 | XP_016860878.1 | 21 | 5860 | |||
XM_017005390.1 | XP_016860879.1 | 20 | 5794 | |||
XM_017005391.1 | XP_016860880.1 | 19 | 5859 | |||
XM_017005392.1 | XP_016860881.1 | 18 | 6028 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK126797.1 | - | 1 | 319 | Chr. 2 - 218755964 - 218759387 [+] on Build GRCh38 |