Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC105379704 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC036479.1 | - | 1 | 2260 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI916626.1 | - | 344 | Chr. - - [-] on Build |