Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYRM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017011084.1 | XP_016866573.1 | 4 | 3184 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK126470.1 | - | 2 | 3028 | Chr. 6 - 5031756 - 5054423 [-] on Build GRCh38 |