Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006712970.3 | XP_006713033.1 | 6 | 4530 | ||
XM_017005718.1 | XP_016861207.1 | 6 | 4530 | |||
XM_017005722.1 | XP_016861211.1 | 5 | 4512 | |||
GenBank mRNA | AF131805.1 | - | 1 | 49 |