Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARAP1-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK125753.1 | - | 2 | 1683 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000500163.2 | - | 2 | 1683 | Chr. 11 - 72700474 - 72705607 [+] on Build GRCh38 |