Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPS15L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017027087.1 | XP_016882576.1 | 25 | 2698 | ||
XM_017027088.1 | XP_016882577.1 | 25 | 2591 | |||
GenBank mRNA | BC028243.1 | - | 1 | 1335 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AW962695.1 | - | 1 | 84 | Chr. 19 - 16356521 - 16357076 [-] on Build GRCh38 |