Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011539270.2 | XP_011537572.1 | 7 | 5531 | ||
XM_011539273.2 | XP_011537575.1 | 5 | 3948 | |||
GenBank mRNA | AK097894.1 | - | 1 | 189 | ||
AK126230.1 | - | 1 | 1095 | |||
AL831850.1 | - | 1 | 110 |