Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | X62009.1 | - | 18 | 2766 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK024353.1 | - | 1 | 1628 | Chr. 5 - 128327657 - 128329772 [-] on Build GRCh38 |