Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPE1P13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC047033.1 | - | 1 | 393 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG122370.1 | - | 1 | 387 | Chr. 2 - 39097991 - 39098686 [-] on Build GRCh38 |