Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5orf42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK022380.1 | - | 1 | 573 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AU122286.2 | - | 1 | 573 | Chr. 5 - 37079097 - 37079942 [-] on Build GRCh38 |