Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCML4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011535703.2 | XP_011534005.1 | 8 | 7092 | ||
XM_011535704.2 | XP_011534006.1 | 8 | 6970 | |||
XM_011535706.1 | XP_011534008.1 | 7 | 6936 | |||
XM_011535709.2 | XP_011534011.1 | 4 | 7126 | |||
XM_011535710.2 | XP_011534012.1 | 6 | 6803 | |||
XM_017010677.1 | XP_016866166.1 | 8 | 7261 | |||
XM_017010678.1 | XP_016866167.1 | 8 | 7174 | |||
XM_017010679.1 | XP_016866168.1 | 8 | 7072 | |||
XM_017010680.1 | XP_016866169.1 | 8 | 7140 | |||
XM_017010681.1 | XP_016866170.1 | 8 | 7050 | |||
XM_017010682.1 | XP_016866171.1 | 8 | 7275 | |||
XM_017010683.1 | XP_016866172.1 | 7 | 6933 | |||
XM_017010685.1 | XP_016866174.1 | 7 | 7060 | |||
XM_017010686.1 | XP_016866175.1 | 7 | 7045 | |||
XM_017010687.1 | XP_016866176.1 | 7 | 6971 | |||
GenBank mRNA | AK128420.1 | - | 1 | 719 |