Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011514680.2 | XP_011512982.1 | 67 | 16921 | ||
XM_011514682.2 | XP_011512984.1 | 66 | 16405 | |||
XM_011514683.2 | XP_011512985.1 | 68 | 15901 | |||
XM_011514684.2 | XP_011512986.1 | 59 | 15687 | |||
XM_011514690.2 | XP_011512992.1 | 32 | 10463 | |||
XM_011514691.2 | XP_011512993.1 | 32 | 16488 | |||
XM_017010944.1 | XP_016866433.1 | 67 | 16403 | |||
XM_017010945.1 | XP_016866434.1 | 66 | 16468 | |||
XM_017010946.1 | XP_016866435.1 | 65 | 16348 | |||
XM_017010947.1 | XP_016866436.1 | 65 | 16279 | |||
XM_017010948.1 | XP_016866437.1 | 58 | 16361 | |||
XM_017010949.1 | XP_016866438.1 | 49 | 15540 | |||
GenBank mRNA | BX538137.1 | - | 1 | 3808 |