Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGTA1P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AF378121.1 | - | LIT2093 alpha1,3GTP1 |
8 | 2889 | |
AF378123.1 | - | LIT2095 alpha1,3GTP3 |
8 | 2938 |