Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017025349.1 | XP_016880838.1 | 9 | 4937 | ||
XM_017025350.1 | XP_016880839.1 | 9 | 4932 | |||
XM_017025352.1 | XP_016880841.1 | 10 | 4105 | |||
XM_017025353.1 | XP_016880842.1 | 9 | 3957 | |||
XM_017025354.1 | XP_016880843.1 | 9 | 4073 | |||
XM_017025355.1 | XP_016880844.1 | 8 | 3925 | |||
XM_017025356.1 | XP_016880845.1 | 9 | 5162 | |||
GenBank mRNA | AK023469.1 | - | 2 | 1054 |