Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC401433 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127717.1 | - | 1 | 2069 |
Gene Symbol: NUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CN258712.1 | - | 1 | 138 |
Gene Symbol: WDR86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011516144.2 | XP_011514446.1 | 7 | 3776 | ||
XM_011516145.2 | XP_011514447.1 | 6 | 3647 | |||
XM_011516147.2 | XP_011514449.1 | 6 | 3576 | |||
XM_011516151.2 | XP_011514453.1 | 5 | 3447 |