Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC401433 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127717.1 | - | 1 | 2007 |
Gene Symbol: NUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AW205479.1 | - | 1 | 64 | ||
CN258712.1 | - | 1 | 76 |
Gene Symbol: WDR86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011516144.2 | XP_011514446.1 | 7 | 3714 | ||
XM_011516145.2 | XP_011514447.1 | 6 | 3585 | |||
XM_011516147.2 | XP_011514449.1 | 6 | 3514 | |||
XM_011516151.2 | XP_011514453.1 | 5 | 3385 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | HY304587.1 | - | 2 | 62 | Chr. 7 - 151377972 - 151404248 [-] on Build GRCh38 |